Would you like to log in again?
Accucore™ XL C18 Drop-in Guard Cartridges, Thermo Scientific
Supplier: Thermo Scientific
Accucore™
74104-013001 74104-012101 74104-014001
10038-688EA 580.45 USD InStock
10038-688 10038-692 10038-690
Accucore™ XL C18 Drop-in Guard Cartridges, Thermo Scientific
Chromatography Columns
New Design protects the main column with minimized impact on retention or efficiency.
  • Economical and Easy To Use
  • Compatible with Uniguard™ Direct-Connection and Stand-Alone Holders
  • Optimum Retention of Non-Polar Compounds
  • Hydrophobic Interaction Mechanism
  • Separates a Broad Range of Analytes
Offer easy, economical protection of the main column with minimized impact on retention or efficiency. Based on Core Enhanced Technology™ using 4 µm solid core particles, Accucore™ XL C18 Drop-in Guard Cartridges allow users of conventional HPLC methods to enjoy performance far beyond that of columns packed with 5, 4, or even 3 µm fully porous particles.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty