Would you like to log in again?
TraceGOLD TG-5MS GC Columns, Thermo Scientific
Supplier: Thermo Scientific
26098-2960 26098-0200 26098-3090 26098-2940 26098-2250 26098-2850 26098-2130 26098-4100 26098-1420 26098-2240 26098-1540 26098-2360 26098-2980 26098-2230 26098-2970 26098-3340 26098-1430 26098-3360 26098-1440 26098-3080 26098-2760 26098-1550 26098-2860 26098-1310 26098-5820 26098-2350 26098-5780 26098-1300
10056-720EA 1234.06 USD InStock
10056-720 10038-938 10038-936 10038-958 10038-956 10038-954 10038-976 10038-984 10038-962 10038-940 10038-960 10038-982 10038-980 10038-968 10038-988 10038-966 10038-944 10038-964 10038-986 10038-974 10038-952 10038-994 10038-950 10038-972 10038-970 10038-992 10038-990 10056-728
TraceGOLD TG-5MS GC Columns, Thermo Scientific
Chromatography Columns
Employ the most widely used MS phase in gas chromatography with the 5% phenyl phase Thermo Scientific TraceGOLD TG-5MS GC Column.
Low polarity phase, 5% diphenyl/95% dimethyl polysiloxane. Low bleed for excellent signal-to-noise ratio, sensitivity and mass spectral integrity. Exceptional inertness ideal for analysis of active compounds. Equivalent to USP G27 phase. Similar to Rxi-5ms, DB-5, HP-5, HP-5ms, Ultra-2, SPB-5, Equity-5, CP-Sil 8. Recommended for semivolatiles, phenols, amines, residual solvents and solvent impurities, drugs of abuse, pesticides, PCB cogeners, and aroclor mixes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty