Would you like to log in again?
RECOM HUMAN PROTEIN IL-2 15.4KDA MW 5UG
Supplier: IBI Scientific
Recombinant human protein IL-2^ Gene ID of 3558. These proteins are lyophilized and are reconstituted with sterile PBS containing at least 0.1% carrier protein. The calculated M.W. of this protein is 15.4kDa. The proteins are expressed in a yeast system and endotoxin free.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
RECOM HUMAN PROTEIN IL-2 15.4KDA MW 5UG 10039-888 RPH-160 Each (5µG) Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty