Would you like to log in again?
Mouse Recombinant GMCSF
Supplier: IBI Scientific
RPM-195 RPM-193 RPM-194 RPM-191 RPM-190 RPM-192 RPM-196
10039-940PK 728.03 USD InStock
10039-940 10039-906 10039-946 10039-902 10039-904 10039-942 10039-944
Mouse Recombinant GMCSF
Proteins and Peptides
IBI recombinant proteins are expressed in a yeast system (Pichia pastoris) which promotes protein folding and post-translational modifications for greater bio-activity. IBI proteins are Endotoxin FREE and are purified without HIS-TAGS or an other TAG. Our yeast expression system has proven to produce more active proteins that more closely resemble their host form.
IBI proteins are lyophilized and can be shipped at room temperature. Proteins can be reconstituted in sterile PBS with at least 0.1% of carrier protein such as BSA or cell assay media.
The recombinant mouse protein GM-CSF has a gene ID of 12981. The calculated molecular weight is 14.1kDa.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty