Would you like to log in again?
RECOM MS PROTEIN MCSF 26.0KDA MW 2X25UG
Supplier: IBI Scientific
Recombinant mouse protein M-CSF (CSF-1)^ Gene ID of 12977. These proteins are lyophilized and are reconstituted with sterile PBS containing at least 0.1% carrier protein. The calculated M.W. of this protein is 26.0kDa. The proteins are expressed in a yeast system and endotoxin free.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
RECOM MS PROTEIN MCSF 26.0KDA MW 2X25UG 10039-918 RPM-252 Pack of 2 (50µµG) Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty