Would you like to log in again?
VWR® Vacuum Filtration Systems, Standard Line
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
FPV103250 FPV403250 FPV413250 FPV203000 FPE414500 FNY412000 SCA217150 FPV113500 FPE404150 SCA207250 SCA207000 FNY412250 FNY202000 SCA407250 FPE204000 SCA407150 FPE414150 FPV113000 FPE404000 FPE204150 FPV203250 SCA407500 FNY212150 SCA207150 FNY402000 FPV103150 SCA407000 FNY412500 FPV213250 FPV403000 FPV413000 SCA417500 FPV403500 FNY212500 FPV203150 SCA217250 SCA207500 SCA417150 FPE404250 FPE214000 FPE214150 FPV103000 FPV403150 FPV103500 FNY402150 SCA417000 SCA217000 FPE214250 FPV213150 FPV213500 FPE414250 FPE404500 FPV113250 FPE414000 FNY212000 SCA417250 FNY202250 FPV203500 FPV413150 FNY412150 FNY202500 FPE204500 FPV213000 FPE214500 SCA217500 FNY202150 FPE204250 FNY402500 FNY402250 FNY212250 FPV413500 FPV113150
76010-420CS 377.98 USD InStock
76010-420 76010-386 76010-422 76010-388 10040-474 76010-382 10040-436 76010-384 10040-438 76008-572 76008-574 76008-600 76008-602 76008-604 76008-606 76008-608 76010-380 10040-472 10040-470 76010-376 10040-464 10040-462 76010-378 76010-410 76010-372 10040-468 76010-374 10040-466 76010-416 76010-418 76010-412 76012-768 76010-414 76008-598 76008-710 76010-370 10040-460 76010-400 76012-770 76010-406 76008-590 76010-408 76008-592 76010-402 76010-368 76008-594 76010-404 76008-596 76008-588 76008-620 76008-622 76008-624 76008-626 76008-708 10040-450 10040-442 76010-398 10040-440 76010-394 10040-446 76010-396 10040-444 10040-448 76008-610 76008-576 76008-612 76008-578 76008-614 76008-616 76010-390 76008-618 76010-392
VWR® Vacuum Filtration Systems, Standard Line
Filters Bottle Top Filters
Systems for vacuum filtration of aqueous solutions including cell culture media, buffers, or other biological fluids.
  • Polystyrene funnel and collection reservoir
  • Each individual unit is lot numbered for traceability
  • Solution can be stored in the collection reservoir until needed
  • Top filter and bottom collector bottle have engraved graduation
  • Designed hose connector can fit multiple hose diameters
PES/PVDF/Nylon/SFCA membrane: Intended for the sterile vacuum filtration of aqueous solutions such as cell culture media and biological fluids.
Ideal for tissue culture applications.
Certifications: ISO 10993-5, 1999. Non pyrogenic. non cytotoxic.
Ordering information: Includes the filtration funnel, vacuum port, lid, collection reservoir, and cap for storage.
Packaging: Individually packaged in easy to peel bags. Receiver bottle cap is individually wrapped.
Order Now
ORDER
COMPLEMENTARY PRODUCTS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty