Would you like to log in again?
Acclaim™ PepMap™ 100 C18 Columns
Supplier: Thermo Scientific
Acclaim™ PepMap™
164570 164199 164940 164213 164942 164943 160428 164944 164946 160434 164535 164572 164941 164569 164534 164945 164537 164560 164568 164212
10040-640EA 511.3 USD InStock
10040-640 10040-652 10040-694 10040-688 10040-644 10040-686 10040-654 10044-938 10040-646 75802-920 10040-658 10818-576 10818-574 10044-940 10818-572 10818-736 10818-734 10040-670 10040-690 10818-732
Acclaim™ PepMap™ 100 C18 Columns
Chromatography Columns
This is the standard column for high-resolution, high-sensitivity protein identification for proteomics and biomarker discovery and verification.
The Thermo Scientific™ Acclaim PepMap column is ideal for high-resolution peptide mapping of tryptic, natural, and synthetic peptides and LC-MS/MS peptide mapping for protein identification, biomarker discovery, and systems biology.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty