Would you like to log in again?
HyPURITY™ ADVANCE™ 3 and 5 µm Columns, Thermo Scientific
Supplier: Thermo Scientific
HyPURITY™
21005-102130 21005-254630 21005-054630 21005-104630 21005-154630
10040-930EA 1279.62 USD InStock
10040-930 10040-942 10045-060 10040-934 10040-938
HyPURITY™ ADVANCE™ 3 and 5 µm Columns, Thermo Scientific
Chromatography Columns
HyPURITY ADVANCE is a polar embedded phase which provides excellent performance for compounds showing poor peak shape with traditional alkyl bonding.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty