Would you like to log in again?
Hypersil GOLD™ Silica HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
Hypersil GOLD™
25105-259070A 25103-014001 25103-153030 25102-152130 25105-254630 25103-154630 25105-154630 25102-102130 25102-202130 25102-052130 25105-254030
10041-154EA 1189.48 USD InStock
10041-154 10041-122 10041-136 10041-158 10041-126 10041-128 10041-118 10041-190 10041-192 10041-184
Hypersil GOLD™ Silica HPLC Columns, Thermo Scientific
Chromatography Columns
Achieve outstanding peak shape and sensitivity with Thermo Scientific™ Hypersil GOLD™ Silica HPLC columns.
The unbonded, highly pure, base-deactivated silica media efficiently separates non-polar and moderately polar organic compounds by normal phase chromatography. Available 1.9 µm particle size columns can improve the speed and efficiency of your applications.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty