Would you like to log in again?
Hypersil GOLD™ C8 HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
25203-154630 25205-054630 25203-012101 25205-154630 25203-011001 25205-104630 25205-100365 25205-254630 25203-254630 25203-104630 25205-011001 25205-109370 25205-012101 25205-259270XL 25205-052130 25205-013001 25205-254030 25205-124030 25203-052130 25205-109070 25202-100365 25203-152130 25203-151030 25202-152130 25205-014001 25205-022135 25203-102130 25202-101030 25203-014001 25205-152130 25205-022130 25202-102130 25203-153030 25203-013001 25202-051030 25203-053030 25205-102130 25203-101030 25202-052130 25203-054630 25203-103030 25203-100365
10045-476EA 837.51 USD InStock
10045-476 10045-490 10045-492 10041-280 10041-282 10041-262 10041-240 10041-284 10041-220 10041-242 10041-222 10041-266 10041-200 10041-202 10041-246 10041-248 10041-226 10041-304 10041-228 10041-208 10045-488 10045-480 10041-290 10045-482 10041-292 10041-270 10045-484 10041-294 10041-272 10041-252 10041-296 10041-254 10041-276 10041-210 10041-298 10041-310 10041-256 10041-212 10041-312 10041-258 10041-216
Hypersil GOLD™ C8 HPLC Columns, Thermo Scientific
Chromatography Columns
For analytes with medium hydrophobicity or when a less hydrophobic phase is required to obtain optimum retention, use Thermo Scientific™ Hypersil GOLD™ C8 HPLC columns for outstanding sensitivity and excellent peak shape.
The selectivity is similar to C18 columns with a predictable elution order but less retention. The lower hydrophobicity allows compounds to elute more quickly, yielding faster separations.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm Int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm Int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm Int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty