Would you like to log in again?
Hypersil GOLD™ Amino HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
25703-154630 25702-152130 25702-102130 25705-014001 25703-013001 25703-014001 25703-153030 25703-152130 25705-254630 25702-202130 25702-052130 25705-254030 25705-154630
10041-682EA 1078.67 USD InStock
10041-682 10041-684 10041-674 10041-620 10041-622 10041-644 10041-612 10041-656 10041-646 10041-602 10041-614 10041-648 10041-608
Hypersil GOLD™ Amino HPLC Columns, Thermo Scientific
Chromatography Columns
Achieve excellent chromatographic separations in weak anion exchange, reversed phase, and normal phase modes with Thermo Scientific™ Hypersil GOLD™ Amino HPLC columns.
These columns, featuring a high-performance aminopropyl phase, provide outstanding peak shape and sensitivity and are available with a 1.9 μm particle size for high speed, high efficiency separations.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm int.Ø require the 1mm UNIGUARD holder (10046-746), 2.1/3 mm int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty