Would you like to log in again?
Hypersil™ BDS C18 HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
Hypersil™
28105-304630 28103-154630 28102-104630 28102-152130 28105-013001 28103-124030 28102-054630 28105-102130 28105-304030 28103-102130 28105-154630 28103-254630 28103-152130 28103-054030 28105-253030 28105-012106 37108-014001 28103-052130 28102-034630 28105-014001 28105-254030 28103-033030 28105-103030 28102-052130 28103-103030 28103-104030 28103-154030 28103-153030 28105-104630 28103-032130 28102-014001 28105-254630 28105-259070A 28103-123030 28103-054630 28103-014001 28105-152130 28105-053030 28105-104030 28105-024006 28105-124030 28105-204630 28105-052130 28105-054630 28102-102130 28105-153030 28102-154630 28105-124630 28103-053030 28103-012101 28105-012101 28103-104630 28103-124630 28105-154030 28103-074630 28103-013001 28105-123030
10045-850EA 969.75 USD InStock
10045-850 10042-082 10045-898 10045-924 10045-848 10045-926 10042-124 10042-122 10042-120 10042-096 10042-094 10045-920 10045-844 10045-922 10042-090 10045-846 10042-138 76332-354 10042-136 10045-836 10045-914 10042-134 10045-838 10045-916 10042-056 10042-130 10042-062 10042-060 10045-830 10045-912 10042-108 10042-106 10045-902 10042-104 10045-826 10042-068 10045-904 10042-146 10045-828 10045-906 10042-144 10042-100 10045-908 10042-074 10045-820 10045-822 10046-210 10042-118 10042-116 10045-814 10042-114 10045-816 10042-112 10045-818 10042-110 10042-078 10042-076
Hypersil™ BDS C18 HPLC Columns, Thermo Scientific
Chromatography Columns
For a robust, general-purpose column, use Thermo Scientific™ Hypersil BDS C18 columns.
These economical columns feature reduced silanol activity and provide exceptional reliability for QA/QC labs. Built on the renowned Hypersil silica backbone, the base-deactivated silica is one of the most popular packing materials available.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm int.Ø require the 1mm UNIGUARD holder (10046-746), 2.1/3 mm int.Ø require the 2/3 mm UNIGUARD Holder (10046-748) and 4/4.6 mm int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty