Would you like to log in again?
Hypersil™ ODS C18 HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
Hypersil™
30103-103030 30103-154630 30105-104630 30105-122130 30103-153030 30103-102130 30103-252130 30103-154030 30103-012101 30105-204030 30105-254630 30105-024006 30103-104030 30105-252130 30103-254030 30110-259070 30105-102130 30105-014001 30105-259070A 30105-304030 30110-014001 30110-304630 30105-052130 30105-022106 30103-152130 30103-124030 30105-103030 30105-154030 30103-034630 30105-202130 30105-259270A 30105-153030 30105-259070 30105-154630 30103-124630 30103-254630 30105-204630 30105-152130 30103-053030 30105-259270 30105-304630 30110-204630 30103-014001 30105-124630 30110-304030 30110-254030 30110-254630 30105-254030 30105-013001 30103-013001 30103-104630 30105-124030 30105-104030 30105-054630 30105-053030 30105-253030 30105-014006 30110-104630 30105-012101 30105-123030 30103-054630 30103-064630
10042-360EA 1106.22 USD InStock
10042-360 10042-448 10042-404 10042-328 10042-446 10042-326 10042-444 10042-400 10042-442 10042-366 10042-440 10042-364 10042-362 10042-370 76332-358 10042-418 76332-356 10042-338 10042-416 10042-458 10042-414 10042-334 10042-378 10042-456 10042-412 10042-376 10042-410 10042-454 10042-374 10042-330 10042-452 10042-450 10042-382 10042-380 10042-428 10042-426 10042-468 10042-424 10042-348 10042-422 10042-346 10042-420 10042-344 10042-388 10042-462 10042-342 10042-460 10042-384 10042-340 10042-392 10042-470 10046-054 10046-052 10042-358 10042-436 10042-356 10042-398 10042-432 10042-474 10042-430 10042-350 10042-472
Hypersil™ ODS C18 HPLC Columns, Thermo Scientific
Chromatography Columns
Classical Hypersil stationary phases have global recognition as an industry standard in HPLC, providing an effective analytical tool.
Hypersil phases are well established and referenced in many HPLC methods worldwide.
Certifications: Manufactured under ISO 9001-2000 quality guidelines.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD Holders. Cartridges with a 1 mm Int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm Int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm Int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty