Would you like to log in again?
Hypersil™ SAS C1 HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
30505-254630 30505-154630 30505-014001 30505-124630 30505-204630
10042-590EA 1008.11 USD InStock
10042-590 10042-588 10042-574 10042-586 10042-594
Hypersil™ SAS C1 HPLC Columns, Thermo Scientific
Chromatography Columns
Classical Hypersil stationary phases have global recognition as an industry standard in HPLC, providing an effective analytical tool.
Hypersil phases are well established and referenced in many HPLC methods worldwide. They are manufactured under ISO 9001-2000 quality guidelines.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm int.Ø require the 1mm UNIGUARD holder (10046-746), 2.1/3 mm int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty