Would you like to log in again?
Hypersil™ APS-2 HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
Hypersil™
30703-124030 30703-014001 30703-102130 30703-254630 30705-259070 30703-012101 30703-054630 30705-014001 37705-254630 30703-152130 30705-254030 30705-103030 30703-154630 30705-154630 30705-304030 30705-254630 30705-259270A
10042-608EA 918.04 USD InStock
10042-608 10042-628 10042-618 10042-626 10042-602 10042-658 10042-634 10042-598 10042-620 10042-642 10042-664 10042-610 10042-662 10042-652 10042-660 76332-360 10046-234
Hypersil™ APS-2 HPLC Columns, Thermo Scientific
Chromatography Columns
Classical Hypersil stationary phases have global recognition as an industry standard in HPLC, providing an effective analytical tool.
Hypersil phases are well established and referenced in many HPLC methods worldwide.
Certifications: Manufactured under ISO 9001-2000 quality guidelines.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm Int.Ø require the 1mm UNIGUARD holder (10046-746), 2.1/3 mm Int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm Int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty