Would you like to log in again?
BetaBasic™ 8 3 and 5 µm Columns, Thermo Scientific
Supplier: Thermo Scientific
BetaBasic™
71405-154630 71405-104630 71405-014001 71403-154630 71405-052130 71405-254630
10043-292EA 1256.55 USD InStock
10043-292 10046-340 10043-358 10043-304 10043-364 10043-352
BetaBasic™ 8 3 and 5 µm Columns, Thermo Scientific
Chromatography Columns
BetaBasic columns are highly efficient and reproducible, and feature a 150 Å pore size for small molecules, peptides and protein digests.
Superb pH stability.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty