Would you like to log in again?
BetaBasic™ 8 Javelin Guard Columns
Supplier: Thermo Scientific
BetaBasic™
71405-022106 71405-012106
10043-306EA 723.1 USD InStock
10043-306 10043-300
BetaBasic™ 8 Javelin Guard Columns
Chromatography Columns
BetaBasic™ 8 Javelin guard columns are a good first choice for compounds typically considered difficult to analyze. It is used for analytical HPLC Column.
  • Has a carbon load of 7%
  • pH 2 to 8
  • L7 USP Type
  • 200 m²/g surface area
  • Not encapped
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty