Would you like to log in again?
ProPac™ SCX Cation Exchange Protein Columns, Thermo Scientific
Supplier: Thermo Scientific
063462 063700 074628 074643 088769 063456 054995
10042-826EA 1998.61 USD InStock
10042-826 10044-502 10044-532 10044-500 10044-328 10043-310 10044-384
ProPac™ SCX Cation Exchange Protein Columns, Thermo Scientific
Chromatography Columns
For separations of proteins and their variants using Thermo Scientific™ ProPac SCX and WCX strong and weak cation-exchange columns, which provide exceptionally high resolution.
Based on stationary phases composed of 10 µm nonporous polymeric beads, these columns resolve isoforms that differ by a single charged residue. A thin, hydrophilic layer grafted to the particle core eliminates unwanted secondary interactions. The cation-exchange surface provides pH-based selectivity control and fast mass transfer for high-efficiency separation and moderate capacity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty