Would you like to log in again?
BioBasic™ 4 HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
72305-154030 72305-052130 72305-051030 72305-014001 72305-152130 72305-101030 72305-154630 72305-254630 72305-104630 72305-012101 72305-252130 72305-102130
10043-604EA 1221.26 USD InStock
10043-604 10043-606 10043-610 10043-632 10043-622 10043-634 10043-596 10043-620 10043-576 10043-618 10043-594 10043-572
BioBasic™ 4 HPLC Columns, Thermo Scientific
Chromatography Columns
For outstanding separation of larger peptides and proteins, choose Thermo Scientific™ BioBasic 4 HPLC Columns.
These high performance columns are designed for the analysis of proteins, peptides and other biomolecules with high hydrophobicity.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD Holders. Cartridges with a 1mm ID require the 1 mm UNIGUARD Holder (10046-746), 2.1/3 mm ID require the 2/3 mm UNIGUARD Holder (10046-748) and 4/4.6 mm ID require the 4/4.6 mm UNIGUARD Holder (10046-744).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty