Would you like to log in again?
Syncronis™ HILIC HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
Syncronis™
97503-052130 97503-154630 97502-052130 97505-014001 97505-052130 97502-102130 97505-153030 97505-154630 97505-102130 97505-152130 97505-254630 97503-104630 97505-012101
10044-180EA 1316.87 USD InStock
10044-180 10044-174 10044-152 10044-150 10044-162 10044-140 10044-134 10044-156 10044-168 10044-146 10044-176 10044-144 10044-188
Syncronis™ HILIC HPLC Columns, Thermo Scientific
Chromatography Columns
Ensure consistent, predictable separations of polar and hydrophilic analytes using Thermo Scientific™ Syncronis HILIC columns.
  • Zwitterion ligand gives neutral, highly polar surface
  • No need for ion-pair reagents or derivatisation
  • Highly pure, high surface area silica
Highly pure, high surface area silica and dense stationary phase coverage minimize secondary interactions and deliver reproducible high-resolution separations.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm Int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm Int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm Int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty