Would you like to log in again?
DNAPac™ PA100 Nucleic Acid Columns, Thermo Scientific
Supplier: Thermo Scientific
DNAPac™
043010 088761 088764 088760 043011 043018 088759 088765
10044-378EA 1022.62 USD InStock
10044-378 10044-316 10044-376 10044-374 10044-320
DNAPac™ PA100 Nucleic Acid Columns, Thermo Scientific
Chromatography Columns
Achieve high-resolution analysis and purification of synthetic oligonucleotides with the Thermo Scientific™ DNAPac™ PA100 column.
This strong anion-exchange column resolves full length oligonucleotides from n–1, n+1, and other failure sequences not possible with other columns. For more complex separations, the polymeric properties of the DNAPac PA100 column allow flexibility to select ideal conditions, including high pH, high temperature, or chaotropic agents.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty