Would you like to log in again?
DNAPac™ PA200 Nucleic Acid Columns, Thermo Scientific
Supplier: Thermo Scientific
088781 063421 062998 063419 063423 088780 063000 063425
10044-488EA 1394.1 USD InStock
10044-488 10044-490 10044-350 10044-494 10044-492 10044-476 10044-352 10044-474
DNAPac™ PA200 Nucleic Acid Columns, Thermo Scientific
Chromatography Columns
For ultra-high-resolution HPLC analysis of synthetic oligonucleotides, use Thermo Scientific™ DNAPac PA200 columns.
These small particle size, strong anion-exchange columns resolve full length oligonucleotides from n–1, n+1, and other failure sequences not possible with other columns. Control the retention times and selectivity by choice of salt, pH, and solvent, and tailor the separation to the requirements of your different oligonucleotide analysis challenges.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty