Would you like to log in again?
MAbPac™ SCX-10 Columns 5 µm, Thermo Scientific
Supplier: Thermo Scientific
088782 078655 078656 085198 088783
10044-356EA 5689.4 USD InStock
10044-356 10044-846 10044-844 10044-922 10044-354
MAbPac™ SCX-10 Columns 5 µm, Thermo Scientific
Chromatography Columns
For high-resolution, high efficiency analysis of monoclonal antibodies (MAbs) and associated variants, use Thermo Scientific™ MAbPac SCX-10 strong cation-exchange columns
The unique nonporous pellicular resin provides the resolving power to separate MAb variants that differ by as little as one charged residue. A hydrophilic layer surrounds the polymeric beads and eliminates hydrophobic interactions with the resin, resulting in very efficient peaks. A proprietary grafted cation-exchange surface provides pH selectivity control for high-resolution separations.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty