Would you like to log in again?
ProPac™ HIC Hydrophobic Interaction Columns, Thermo Scientific
Supplier: Thermo Scientific
ProPac™
063655 063665 063653 074197
10044-522EA 2191.58 USD InStock
10044-522 10040-598 10044-518 10044-520
ProPac™ HIC Hydrophobic Interaction Columns, Thermo Scientific
Chromatography Columns
Achieve high-resolution separation of proteins and peptides under non-denaturing conditions with Thermo Scientific™ ProPac HIC-10 hydrophobic interaction chromatography columns.
Developed with multi-functional ligand attachments, these silica-based columns provide exceptional hydrolytic stability under the highly aqueous conditions used in HIC.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty