Would you like to log in again?
Acclaim™ Mixed-Mode WAX-1 LC Columns, Thermo Scientific
Supplier: Thermo Scientific
069686 064984 069704 067084 064985 088785 071908 071909 070089 070088
10044-568EA 1437.96 USD InStock
10044-568 10044-722 10044-566 10044-720 10040-538 10044-680 10044-618 10044-652 10852-494 10044-682
Acclaim™ Mixed-Mode WAX-1 LC Columns, Thermo Scientific
Chromatography Columns
When you need powerful selectivity control for the separation of anionic molecules, use Thermo Scientific™ Acclaim Mixed-Mode WAX-1 LC columns.
The silica-based stationary phase features a hydrophobic alkyl chain with an ionizable terminus that provides both hydrophobic reversed-phase and weak anion-exchange properties. This unique chemistry produces a multimode separation mechanism for simultaneous separation of acidic, basic, and neutral molecules. The selectivity is fully adjustable through changes to ionic strength, pH, or organic solvent content.
Ordering information: Guard Cartridges require use of the corresponding guard holder. See Acclaim Guard Kit Holder and Coupler V-2 (Part No. 10046-750).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty