Would you like to log in again?
Acclaim™ Organic Acid (OA) LC Columns, Thermo Scientific
Supplier: Thermo Scientific
070087 062903 069700 070086 062902 071987
10044-644EA 276.79 USD InStock
10044-644 10044-754 10044-678 10044-676 10044-472 10044-470
Acclaim™ Organic Acid (OA) LC Columns, Thermo Scientific
Chromatography Columns
For unparalleled performance in separating hydrophilic, aliphatic, and aromatic organic acids, use Thermo Scientific™ Acclaim OA reversed-phase silica columns.
These high-efficiency columns are compatible with 100% aqueous mobile phases and have excellent hydrolytic stability at low pHs for excellent selectivity in the separation of organic acids. You'll achieve high-resolution separations with fast run times and minimal sample preparation using Acclaim OA columns for the analysis and quality testing of foods, beverages, pharmaceuticals, chemical intermediates, and environmental samples.
Ordering information: Guard Cartridges require use of the corresponding guard holder. See Acclaim Guard Kit Holder and Coupler V-2 (VWR Part No. 10046-750).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty