Would you like to log in again?
Acclaim™ Surfactant Plus LC Columns, Thermo Scientific
Supplier: Thermo Scientific
078960 078953 078959 078952 078955 078950 078951 078954
10044-864EA 1383.38 USD InStock
10044-864 10044-862 10044-874 10044-868 10043-336 10044-866 10044-872 10044-870
Acclaim™ Surfactant Plus LC Columns, Thermo Scientific
Chromatography Columns
Acclaim Surfactant Plus is a new generation of columns offering improved performance and higher throughput for analyzing surfactants.
They have similar selectivity to the Acclaim Surfactant columns, but have been designed with exceptionally low bleed for use with charged aerosol detectors (CAD) and mass spectrometers (MS). These columns can be used to separate a wide variety of surfactants including anionic, cationic, nonionic and amphoteric surfactants. Additionally, these columns can be used with evaporative light scattering detectors (ELSD), suppressed conductivity detectors (SCD), and UV-Vis detectors (UV).
Ordering information: Guard Cartridges require use of the corresponding guard holder. See Acclaim Guard Kit Holder and Coupler V-2 (Part No. 10046-750).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty