Would you like to log in again?
Acclaim™ SEC LC Columns, Thermo Scientific
Supplier: Thermo Scientific
079722 082739 079726 079724 079721 079723 082740 079725
10044-886EA 3288.08 USD InStock
10044-886 10044-876 10044-920 10044-884 10044-878 10044-882 10044-918 10044-880
Acclaim™ SEC LC Columns, Thermo Scientific
Chromatography Columns
Separate water-soluble polymers and oligomers with Thermo Scientific™ Acclaim™ SEC LC Columns.
These resin-based, high-performance, size-exclusion chromatography columns come in two sizes to cover a broad molecular weight range: 5 µm, 300Å for the SEC-300 column and 7 µm, 1000Å for the SEC-1000 column. The combination of a proprietary mono-dispersed multi-pore hydrophilic resin and small particle size provides high-resolution and good linearity of the calibration curve with no inflection points for accurate, reliable analysis of a variety of water-soluble polymers.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty