Would you like to log in again?
Hypersil GOLD™ Phenyl HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
25905-259070A 25903-013001 25905-154630 25905-102130 25903-104630 25903-012101 25902-152130 25903-154630 25903-014001 25903-102130 25905-254630 25902-102130 25902-052130 25905-012101
10045-762EA 1161.35 USD InStock
10045-762 10045-730 10045-756 10045-734 10045-746 10041-780 10041-792 10041-770 10041-776 10041-788 10041-766 10041-778 76332-252 10041-758
Hypersil GOLD™ Phenyl HPLC Columns, Thermo Scientific
Chromatography Columns
Get excellent retention and unique selectivity for aromatic and moderately polar analytes with Thermo Scientific™ Hypersil GOLD™ Phenyl reversed phase HPLC columns.
A butyl (C4) linker in the phenyl bonded phase allows for superior alignment of the phenyl ring with aromatic molecules, thus enhancing pi-pi interactions and improving the retention of the molecules. The C4 linker also provides the stationary phase with moderate hydrophobicity, making it ideal for the separation of analyte mixtures with varying polarity and aromaticity. The outstanding peak shape results in greater sensitivity.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD Holders. Cartridges with a 1 mm int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty