Would you like to log in again?
Idex® High-Purity Plus PFA Tubings
Supplier: Antylia Scientific
Idex®
02012-80 02012-86 02012-82 02012-84
100452-488EA 240.25 USD InStock
100452-488 100457-898 100452-494 100452-508
Idex® High-Purity Plus PFA Tubings
Tubing Chromatography Tubing
Excellent stress cracking resistance even when exposed to aggressive fluorosurfactants.
  • Low gas permeability and outstanding resistance to solvents
  • Manufactured from premium-grade PFA
  • Maximum operating temperature 80 °C
High purity plus PFA tubing withstands repeated flexing and offers high continuous service temperature (recommended max operating temperature 80 °C). Other added benefits include clearer walls, smooth surface texture, and excellent polymer purity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty