Would you like to log in again?
Javelin™ Direct-Connection Guard Columns, Thermo Scientific
Supplier: Thermo Scientific
Javelin™
70105-012106 70105-022106 71505-014006 71505-024006 71505-022106
10046-396EA 719.42 USD InStock
10046-396 10046-394 10043-166 10046-402 10043-154
Javelin™ Direct-Connection Guard Columns, Thermo Scientific
Chromatography Columns
Javelin guard columns provide convenient, easy-to-use protection for analytical HPLC columns.
An economical means to extend column lifetime and improve performance, the guard columns feature a direct-connection, fingertight design for convenience and maximum efficiency. The patented design requires no holder, allowing quick and easy replacement. For all popular Thermo Scientific stationary phases.
The patented Javelin design has a ¹/₁₆" CPI tip that attaches directly to the inlet of the HPLC column, without the need for tubing or wrenches. To replace, simply remove and discard the entire guard column for fast, easy change over. Javelin guard columns come in all popular Thermo Scientific phases and are available in 10 mm lengths in 4.0, 2.1, 3.0 and 1.0 mm Int.∅. The 4.0 mm Int.∅ Javelin guard is designed for use with both 4.6 and 4.0 mm Int.∅ columns. Javelin guard columns are provided in convenient packs of 4 columns, with the exception of Hypercarb which is supplied in packs of 2. Javelin guard columns with inlet tips for Waters columns are readily available upon request. Javelin guard columns are also popular as analytical columns although they are provided without QC test results. For short, fast columns for LC/MS and high throughput applications, including direct-connection columns and fully labeled and traceable columns, please refer to Javelin HTS columns and DASH HTS columns.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty