Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
HyperSep Glass Block Manifolds, Thermo Scientific
Supplier: Thermo Scientific
60104-233 60104-232
10047-016EA 2684.62 USD InStock
10047-016 10047-014
HyperSep Glass Block Manifolds, Thermo Scientific
Manifolds
Provides multiple processing of SPE or SLE cartridges to improve throughput and reproducibility.
Delivery information: Includes glass block manifold lid, cover gasket, vacuum gauge, valve assembly, tips (16 or 24), adjustable collection rack, bulkhead luer fittings, plugs (16 or 24) and manifold safety tray.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty