Would you like to log in again?
HyperSep™ Aminopropyl SPE Cartridges
Supplier: Thermo Scientific
HyperSep™
60108-424 60108-425 60108-518 60108-739 60108-738 60108-519 60108-432 60108-740 60108-364
10047-294EA 343.48 USD InStock
10047-294 10047-292 10047-314 10048-112 10047-302 10047-236 10047-316 10048-116 10048-114
HyperSep™ Aminopropyl SPE Cartridges
Chromatography Columns
A polar sorbent for both polar and anion exchange interactions.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty