Would you like to log in again?
Neurotransmitter Transporter Uptake Assay Kit, Molecular Devices
Supplier: Molecular Devices
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
R6138 R6138 R8173 R8174 R8174 R8173
10048-836EA 5286.05 USD InStock
CA10048-834 10048-836 CA10048-832 10048-834 CA10048-836 10048-832
Neurotransmitter Transporter Uptake Assay Kit, Molecular Devices
Assays Cellular Assays
Neurotransmitter transporters play a key role in depression and neurodegenerative diseases (NDD) such as Alzheimer's and Parkinson's.
  • Replaces radioactive assays
  • One-step, homogeneous assay format
  • Two assay setup
The ability to monitor serotonin, norepinephrine and dopamine neurotransmitter uptake is key to a better understanding of these diseases.
Homogeneous, live-cell fluorescent assay replaces radioactive assays, reducing disposal costs and eliminating safety risks associated with radiolabeled assays. One-step, homogeneous assay format is amenable to automation and high-throughput screening. Two assay setup options offer greater flexibility: endpoint mode for high-throughput applications, or real-time kinetic mode for mechanistic studies.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty