Would you like to log in again?
Anionic Gold Tracer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
25470 25473 25460 25466 25469 25463 25472 25467 25461 25464
100487-886EA 343.72 USD InStock
100487-886 100487-896 100487-884 100487-894 100487-900 100487-888 100487-898 100487-882 100487-892 100487-890
Anionic Gold Tracer, Electron Microscopy Sciences
Protein Linking and Labeling Systems
Anionic tracers are used to visualize positive charged moieties in specimens
  • Particle sizes of 0 8, 6, 10, 15 and 25 nm
  • Guaranteed shelf life of 18 months
  • Supplied in PBS with 15 mM NaN3
Aurion Gold Tracers are used to visualize charged moieties in specimens: anionic tracers bind to polycationic moieties (basic proteins e.g. histones), cationic tracers bind to polyanionic moieties (membranes, acidic proteins).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty