Would you like to log in again?
Gold Sol, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
25516 25509 25514 25510 25512
100487-930EA 399.66 USD InStock
100487-930 100487-926 100487-924 102091-022 100487-928
Gold Sol, Electron Microscopy Sciences
Protein Linking and Labeling Systems
Solutions of high quality unconjugated gold particles.
  • Particle sizes of 0.8, 6, 10 ,15 and 25 nm
  • Guaranteed shelf life of 12 months
  • Monodisperse particle population
To prepare a high quality (immuno) gold conjugate it is important to have particles with uniform size and highly adsorptive surfaces.
Introduction
The preparation of conventional gold reagents is based on gold particles with a diameter suited for direct electron microscopic visualization. Aurion offers a range of these Reagents which cover most of the approaches in transmission and scanning electron microscopy.
Aurion Gold Sols (solutions of high quality unconjugated gold particles) provide opportunities for users to prepare conjugates with primary antibodies, ligands and other binding agents with the same particle characteristics as in the Conventional Immunogold Reagents.
Product Description
AURION Gold Sols are prepared according to a unique protocol, warranting narrow size distribution and reproducible adsorption characteristics.
The AURION gold sols are available in the same size range as the Conventional Immunogold Reagents: 6, 10, 15 and 25 nm. The particle population is monodispersed and thus shows minimal size variation and overlap. Typically, the coefficient of variance for the 6 and 25 nm particle size sols is less than 12%, whereas the 10 and 15 nm size sols show less than 10% variation. Actual lot specifications (size, variation and expiry date) are indicated on the accompanying package insert.
Package size: 100 ml of high quality gold sol at an OD520 nm of approximately 1.
Storage
AURION gold sols have a guaranteed shelf life of 12 months from the date of quality control analysis.
The products should be stored at 4 – 8°C.
Freezing is not recommended.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty