Would you like to log in again?
Col-Aurion Total Protein Stain, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
25536
100487-932EA 266.45 USD InStock
100487-932
Col-Aurion Total Protein Stain, Electron Microscopy Sciences
Protein Linking and Labeling Systems
Col-Aurion is a total protein stain consisting of a solution of coated gold particles with an average particle diameter of 15 nm. The staining principle is based on the electrostatic binding of negatively charged gold particles to proteins with a positive charge present at low pH.
  • pH 3.2
  • Ready to use
Col-Aurion is developed for the sensitive staining of electrophoretically separated protein bands on nitrocellulose or PVDF™ blotting membranes. Colloidal gold particles accumulate at the site of the protein bands on the membrane, generating an intense dark red staining pattern. The total protein stain assists in assessing immunoblotting results and to evaluate the effectivity of the blotting procedure.
The negative surface charge of colloidal gold particles is responsible for their high affinity for positively charged macromolecules. This characteristic was the basis for the development of a total protein stain based on colloidal gold.
A unique feature of Col-Aurion is the use of BSA-c™ to shield off the surface of the gold particles. Destabilization of gold particles that might occur as a result of interaction with detached protein is thus prevented. The strong negative charge of BSA-c™ gives an additional increase in sensitivity of the stain.
Delivery information: For removal of surplus of weakly bound protein from the membrane, a 10 ml vial of Tween-20™ is included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty