Would you like to log in again?
AURION BSA-c™ Aurion Incubation Solution Additive, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
25558 25557
100487-938EA 444.97 USD InStock
100487-938 100487-936
AURION BSA-c™ Aurion Incubation Solution Additive, Electron Microscopy Sciences
Buffers Microscopy Buffers
Aurion incubation solution additive AURION BSA-c™, unique incubation buffers additive with an unparalleled ability to effectively prevent charge-based background. BSA-c™ are prepared by acetylation of bovine serum albumin (BSA). Polycationic sites in the specimen interact readily with negatively charged acetylated BSA molecules. This significantly reduces the risk that such sites might bind negatively-charged immunoreagents and immunogold conjugates and thus reduces the risk of background.
AURION BSA-c™ buffers suppresses background competitively with little or no effect on the specific reaction. Its successful application is not limited to immunogold detections, but it is equally efficient in fluorescent and enzyme-based detection systems. AURION BSA-c™ concentrations as low as 0.01 to 0.1% inhibit binding of gold conjugate to polycationic poly-l-lysine coated grids almost completely (>99%).
In the blocking step, hydrophobic moieties causing stickiness in the specimen surface are rendered hydrophilic to minimize background. The more dynamic charge-based interaction between the specimen surface and immunoreagents also needs to be controlled to eliminate background.
The surface properties of the specimen can be simplified by division into four compartments. They are negatively charged (polyanions, proteins, especially after aldehyde fixation, lipids), neutral (0) and positively charged (histone proteins, polycations) and H hydrophobic (lipids, fat droplets, resins). After an appropriate blocking step these areas are covered with blocking compounds.
In low ionic strength media negatively charged antibodies and gold conjugates are repulsed by negatively charged specimen areas which frequently may contain the antigens to be detected. Background does not likely occur in such areas. The positively charged areas attract antibodies and gold conjugates potentially leading to background. In a moderate ionic strength incubation solution, repulsion and attraction are diminished due to the presence of ions. The negatively charged BSA-c™ competes with the negatively charged antibodies and gold reagents for non-specific binding to the positively charged specimen compounds, thus reducing background to the greatest possible extent without interfering with antigen detection.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty