Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Mouse Monoclonal Antibody (Gold, >1 nm (ultra-small))

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

25581 25580
100487-956EA 448.62 USD InStock
100487-956 100487-954
Anti-IgG Mouse Monoclonal Antibody (Gold, >1 nm (ultra-small))
Antibodies
Mouse Monoclonal anti-FITC Gold Conjugates For the electron microscopic visualization of FITC labels used in light microscopic specimens. For retrieval of faded FITC in DNA/RNA probes, fatty acids, and proteins.

For the electron microscopic visualization of FITC labels used in light microscopic specimens. For retrieval of faded FITC in DNA/RNA probes, fatty acids, and proteins.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: Gold, >1 nm (ultra-small)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty