Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 

Anti-IgG Donkey Polyclonal Antibody (Gold, 10 nm)

Supplier: Electron Microscopy Sciences
25814 25815
100488-040EA 634.35 USD InStock
100488-040 100488-042
Anti-IgG Donkey Polyclonal Antibody (Gold, 10 nm)
Antibodies
Secondary ImmunoGold conjugates are based on antibodies raised in Donkey. It is used for double labeling experiment in combination with, for instance a mouse primary antibody, since applicable secondary antibodies are either raised in rabbit or goat. Ultra small conjugates have far less influence on the adsorbed antibodies or detecting molecules, thus behave as though they are uncoupled. Used with R-Gent SE-LM and SE-EM silver enhancement reagents they are the best choice for any application.

It is impossible to perform a double labeling experiment in combination with, for instance a mouse primary antibody, since applicable secondary antibodies are either raised in rabbit or goat. To overcome this difficulty the Aurion Series of Secondary ImmunoGold conjugates is based on antibodies raised in Donkey, including Donkey-anti-Goat. It is then known that mixing Donkey-anti-Goat and Donkey-anti-Mouse provides a successful solution. Ultra small conjugates have far less influence on the adsorbed antibodies or detecting molecules, thus behave as though they are uncoupled. Used with R-Gent SE-LM and SE-EM silver enhancement reagents they are the best choice for any application.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Gold, <1 nm (ultra-small)
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty