Would you like to log in again?
Kodak T-MAX 400 Professional Films, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
KODAK®
74170 74166 74168
100488-108RL 280.79 USD InStock
100488-108 100488-110 100502-684
Kodak T-MAX 400 Professional Films, Electron Microscopy Sciences
Photographic Film
Especially useful for dimly lighted subjects, fast action, extending the distance range, and flash pictures. Used in scientific and biomedical work, especially where fluorescence photography is required.
  • High speed (EI 400)
  • Very high sharpness; extremely fine grain, high resolving power
  • High degree of enlargement
  • Shorter push-processing development time
  • Total darkness
Recommended kodak developers: T-Max, D-76, HR-110, Microdol-X.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty