Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Cheese Cloth, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71748-00
100488-116RL 141.61 USD InStock
100488-116
Cheese Cloth, Electron Microscopy Sciences
Wipes Reusable Cleaning Cloths Cloths for General Applications
Cotton woven cloth is suitable for cleaning and polishing.
  • (44×36") 44 lines/inch horizontal
  • 36 lines/inch vertical
  • Supplied in 60 yard rolls 36" in width
This configuration allows to cut out the exact length desired.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty