Would you like to log in again?
Capsules, Gelatin, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
70102 70100 70110 70105 70104 70101 70103
100488-366PK 57.68 USD InStock
100488-366 100488-368 100488-356 100488-358 100488-360 100488-362 100488-364
Capsules, Gelatin, Electron Microscopy Sciences
Moulds Moulds for Histology
Natural, hard gelatin capsules are used as an embedding mold for water miscible resins, or resins which need to be cured by transmitted light. The gelatin capsules dissolve or disintegrate within 5 minutes when immersed in a 0.5% HCl solution at 36 to 38 °C, within 2 minutes in water at 37 °C or in artificial gastric juice.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty