Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cambridge S-4, Mark II, S-410 SEM Mounts, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

75070 76181 76180 75060 75050
100488-640PK 174.22 USD InStock
100488-640 100488-642 100488-776 100488-778 100488-638
Cambridge S-4, Mark II, S-410 SEM Mounts, Electron Microscopy Sciences
Specimen Mounting Supplies
Surface: 31.7 mm (1 1/4") in diameter and 6.4 mm (3/16”).

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty