Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

45° Angle SEM Aluminum Mounts, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

75342 75241-30 75240 75241-60 75241-40 75340 75241-10 75241-50 75241-20 75242
100488-696PK 537.41 USD InStock
100488-696 100488-678 102098-158 102098-156 102098-154 102098-152 102098-150 102098-160 100488-680 100488-694
45° Angle SEM Aluminum Mounts, Electron Microscopy Sciences
Specimen Mounting Supplies
1) Surface measures 12.7 mm (1/2") in diameter and has 3.1 mm (1/8") pin
2) Surface measures 10 mm (3/8") in diameter and height
3) Head measures 15 mm (5/8") in diameter, Height: 10 mm (3/8")
4) Head measures 25 mm (1") in diameter, Height: 20 mm (13/16")

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty