Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Easy-Load Slotted Cassettes for Automated Labeling Instruments, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70060-T 70062-PE 70063-Y 70060-W 70063-B 70062-A 70060-P 70061-G 70062-G 70063-GR 70061-A 70062-P 70061-GR 70061-O 70062-L 70061-P 70062-W 70062-B 70063-PE 70063-T 70061-T 70060-B 70060-PE 70062-Y 70063-O 70060-L 70062-T 70060-GR 70061-PE 70060-A 70063-L 70062-GR 70061-B 70060-Y 70063-A 70063-W 70060-O 70063-P 70062-O 70061-Y 70061-W 70063-G 70060-G 70061-L
100489-010PK 509.68 USD InStock
100489-010 100489-032 100489-030 100489-018 100489-016 100499-978 100489-014 100489-034 100489-012 100488-982 100499-974 100488-984 100499-976 100488-986 100499-970 100499-972 100488-988 100488-980 100489-000 100489-022 100489-020 100489-008 100488-978 100489-006 100489-028 100489-004 100489-026 100499-968 100489-024 100489-002 100488-992 100488-970 100499-986 100488-994 100499-966 100488-972 100499-982 100488-974 100488-996 100499-984 100488-976 100488-998 100499-980 100488-990
Easy-Load Slotted Cassettes for Automated Labeling Instruments, Electron Microscopy Sciences
Embedding Cassettes Cassettes for Labeling Instruments
The anterior printing area is a 45° angle which suits most cassette labeling instruments
Made from acetal copolymer – This newly patented cassette, allows you to load the Micro-Writer® and Shur/Mark® Cassette Labeling Instruments in one simple operation. Each convenient stack contains 50 unique cassettes. No more manual insertion one-by-one. Save time and money. Two types of cassettes are offered in order to suit your needs: Slotted tissue cassettes and meshed biopsy cassettes.
The anterior printing area is a 45° angle which suits most cassette labeling instruments. Lids are available as one-piece disposable plastic. It can be opened and closed as often as necessary and it always relocks securely without danger of specimen loss. Cassettes are packaged in stacks of 50. Each case contains 2 boxes of 20 stacks. Order cassette bases and cover separately.
Lids are available as one-piece disposable plastic. It can be opened and closed as often as necessary and it always relocks securely without danger of specimen loss. Cassettes are packaged in stacks of 50. Each case contains 2 boxes of 20 stacks. Order cassette bases and cover separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty