Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Secure-Seal™ Hybridization Chambers, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

SecureSeal™
70333-37 70333-10 70333-22 70333-74 70333-01 70333-86 70333-46 70333-44 70333-66 70333-20 70333-32 70333-70 70333-58 70333-30 70333-27 70333-47 70333-40 70333-62 70333-82 70333-76 70333-04 70333-42 70333-34 70333-50 70333-36 70333-83 70333-03 70333-02
102096-680PK 402.43 USD InStock
102096-680 100489-054 100489-052 102096-684 102096-662 100489-050 102096-682 100501-878 100489-058 100489-056 102096-676 102096-674 102096-678 100489-044 102096-690 100489-042 100501-208 102096-672 100489-060 102096-692 102096-670 100489-048 100489-046 102096-688 102096-666 102096-686 102096-664 102096-668
Secure-Seal™ Hybridization Chambers, Electron Microscopy Sciences
Slides Microarray Slides
Design minimize friction, promotes reagent mixing, and facilitates uniform hybridization.

  • Sealable access ports (1.5 mm) in the chamber surface allow for the addition and removal of reactants
  • Adhesive seal tabs create leak-proof chambers that are temperature resistant
  • SecureSeal™adhesive bonds chambers to glass in seconds and removes cleanly and easily even after heating
  • SecureSeals™are ideally suited for protocols utilizing autoradiographic, fluorescent or cheminescent end points
  • Polycarbonate HybriSlip used, 0.25 mm thick

SecureSeal™ adhesive chambers form peel-and-stick enclosures to isolate specimens affixed to glass microscope slides. RNase free, gasketed chambers provide optimum surface-to-volume fluid dynamics for hybridization assays on large or multiple specimens and genomic arrays on glass slides or film coated slides.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty