Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
CultureWell™ Chambered Coverglass, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70460-2R1 70460-50R 70460-3S 70460-4R 70460-8R 70460-2R2
100489-154PK 342.35 USD InStock
100489-154 100489-152 100489-150 100501-886 100489-148 100489-146
CultureWell™ Chambered Coverglass, Electron Microscopy Sciences
Cover Glasses
Chambered coverglass products are optimal for high resolution microscopy.
  • The thin flat coverglass is the optimal platform for high resolution microscopy and high- sensitivity, quantitative, fluorescent imaging
  • Leak-proof design eliminates well-to-well cross contamination
  • Gaskets stabilize coverglass and provide convenient handling using  conventional microscope slide holders and accessories
  • Gaskets are removable for high resolution imaging
  • Gasket may be re-sterilized and reused
  • Well spacing facilitates liquid handling utilizing multichannel pipettes and robotic dispensers
Chambered coverglass products consists of removable / reusable non-cytotoxic silicone gaskets secured to 24 x 50 mm, #1.5 German coverglass. They may be placed in sterile culture plates or 100 mm diameter culture dishes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty