Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
CultureWell™ Chambered Coverglass Plate Inserts, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70461-3S 70461-4R 70461-50R 70461-8R 70461-2R1 70461-2R2
100489-164PK 1669.49 USD InStock
100489-164 100489-162 100489-160 100501-888 100489-158 100489-156
CultureWell™ Chambered Coverglass Plate Inserts, Electron Microscopy Sciences
Cover Glasses
Each CultureWell™ insert is comprised of four chambered coverglass assembled in a disposable frame which is placed in a standard 86 x 128 mm culture plate.
All inserts are sterile and ready to use. The stabilizing insert design and accessory hardware allow fragile coverglass to be easily integrated into protocols for high throughput handling of microscope slides and SBS compliant microtiter plates.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty