Would you like to log in again?
Titer-Tops, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70538-05 70538-10
100489-186PK 61.75 USD InStock
100489-186 100489-188
Titer-Tops, Electron Microscopy Sciences
Seals Microplate Seals
Titer-Tops prevents sample evaporation, well to well contamination, external contamination and sample spillage. Titer-Tops are made from 4.4 mil polyethylene film with an FDA approved non cytotoxic acrylic adhesive.
  • Titer-Tops are chemically resistant and thermally stable
  • Titer-Tops are gamma sterile
  • They are easily pierced with a micropipette tip to provide access to sample wells
Each pre-cut rectangular sheet is 6 x 3.25" with a paper liner. The center section of the liner measures 5 x 3.25" and once it is peeled off it leaves two hand-grip extension tabs on the two ends. These tabs allow for easy application of the Titer-Top to the plate. The acrylic adhesive is waterproof, solvent resistant, and compatible with all plastic microtiter plates. Titer-Tops will stay in place even when exposed to 100% humidity and temperatures ranging from -40 to 100 °C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty